ENA370563399 Enamine

2-(chloromethyl)-5-fluoro-1,3-benzoxazole

  • MDL number MFCD10696613
  • Cas Number : 143708-35-4
  • Molecular Weight : 185.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-105821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105821
Availability details for
Quantity
  • Availability
  • Add to Cart