ENA370563402 Enamine

6-hydrazinopyridazine-3-carboxamide

  • MDL number MFCD00867322
  • Cas Number : 3614-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 251...253
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105824

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105824
Availability details for
Quantity
  • Availability
  • Add to Cart