ENA370563418 Enamine

1-(2-fluorophenyl)-3-methyl-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD11642988
  • Cas Number : 1098352-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-105852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105852
Availability details for
Quantity
  • Availability
  • Add to Cart