ENA370563424 Enamine

1-(butan-2-yl)-1H-imidazole

  • MDL number MFCD06421461
  • Cas Number : 20075-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-105859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105859
Availability details for
Quantity
  • Availability
  • Add to Cart