ENA370563426 Enamine

N-cyclopentyl-4-fluoroaniline

  • MDL number MFCD11145035
  • Cas Number : 557799-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-105861

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105861
Availability details for
Quantity
  • Availability
  • Add to Cart