ENA370563427 Enamine

2-[(4-hydroxyphenyl)sulfanyl]acetic acid

  • MDL number MFCD12784660
  • Cas Number : 42580-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105862

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105862
Availability details for
Quantity
  • Availability
  • Add to Cart