ENA370563428 Enamine

1-(2-methylpropyl)cyclopentane-1-carboxylic acid

  • MDL number MFCD16860494
  • Cas Number : 1210879-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104169

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104169
Availability details for
Quantity
  • Availability
  • Add to Cart