ENA370563438 Enamine

4-[(2-chloropyridin-4-yl)methyl]piperazin-2-one

  • MDL number MFCD16758281
  • Cas Number : 1248482-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105881
Availability details for
Quantity
  • Availability
  • Add to Cart