ENA370563439 Enamine

5-bromo-2-methoxy-N-(pyridin-3-ylmethyl)aniline

  • MDL number MFCD13474144
  • Cas Number : 1306066-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105882

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105882
Availability details for
Quantity
  • Availability
  • Add to Cart