ENA370563442 Enamine

2-ethyl-4,5,6,7-tetrahydro-1,3-benzothiazol-7-one

  • MDL number MFCD16671129
  • Cas Number : 1247386-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105914

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105914
Availability details for
Quantity
  • Availability
  • Add to Cart