ENA370563445 Enamine

2-(propan-2-yl)heptanoic acid

  • MDL number MFCD12794597
  • Cas Number : 116988-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104173

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104173
Availability details for
Quantity
  • Availability
  • Add to Cart