ENA370563447 Enamine

2-(1-methyl-1H-pyrazol-4-yl)imidazo[1,2-a]pyridin-3-amine

  • MDL number MFCD18848206
  • Cas Number : 1338985-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105922

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105922
Availability details for
Quantity
  • Availability
  • Add to Cart