ENA370563448 Enamine

3-methyl-1-(morpholin-4-yl)butan-2-amine

  • MDL number MFCD11154922
  • Cas Number : 1021235-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104175

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104175
Availability details for
Quantity
  • Availability
  • Add to Cart