ENA370563455 Enamine

2-ethenyl-5-methylpyridine

  • MDL number MFCD08703184
  • Cas Number : 3883-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105933
Availability details for
Quantity
  • Availability
  • Add to Cart