ENA370563463 Enamine

1-[2-(2-bromophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-amine

  • MDL number MFCD14615084
  • Cas Number : 1249254-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105942

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105942
Availability details for
Quantity
  • Availability
  • Add to Cart