ENA370563468 Enamine

4-(oxolan-3-ylmethoxy)piperidine

  • MDL number MFCD06248803
  • Cas Number : 883537-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104183

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104183
Availability details for
Quantity
  • Availability
  • Add to Cart