ENA370563473 Enamine

{4-methyl-2-[(4-methylpentan-2-yl)oxy]phenyl}methanamine

  • MDL number MFCD16731729
  • Cas Number : 1249037-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105985

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105985
Availability details for
Quantity
  • Availability
  • Add to Cart