ENA370563474 Enamine

3-fluoro-5-methylbenzene-1-sulfonamide

  • MDL number MFCD21785000
  • Cas Number : 1384429-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105988

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105988
Availability details for
Quantity
  • Availability
  • Add to Cart