ENA370563479 Enamine

4-amino-5-methyl-2-(propan-2-yl)phenol hydrochloride

  • MDL number MFCD00035504

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254...256
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104187

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-104187
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart