ENA370563498 Enamine

5-(difluoromethoxy)-1H-indole

  • MDL number MFCD20662845
  • Cas Number : 947380-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106030
Availability details for
Quantity
  • Availability
  • Add to Cart