ENA370563505 Enamine

2-(1-chloroethyl)-5-(2,4-dichlorophenyl)-1,3,4-oxadiazole

  • MDL number MFCD12187264
  • Cas Number : 1156191-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106049

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106049
Availability details for
Quantity
  • Availability
  • Add to Cart