ENA370563506 Enamine

3-(pyridin-4-yl)but-2-enoic acid

  • MDL number MFCD11172847
  • Cas Number : 103441-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106051

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106051
Availability details for
Quantity
  • Availability
  • Add to Cart