ENA370563509 Enamine

2-(pyrazin-2-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD14615201
  • Cas Number : 1250594-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104259

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104259
Availability details for
Quantity
  • Availability
  • Add to Cart