ENA370563511 Enamine

2-(3,4-dimethoxyphenyl)-2-methylpropanoic acid

  • MDL number MFCD11036939
  • Cas Number : 59212-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84...86
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106056
Availability details for
Quantity
  • Availability
  • Add to Cart