ENA370563525 Enamine

3-ethyl-1,2,4-thiadiazole-5-thiol

  • MDL number MFCD21186265
  • Cas Number : 63688-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104261

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104261
Availability details for
Quantity
  • Availability
  • Add to Cart