ENA370563526 Enamine

N'-hydroxy-1-benzofuran-2-carboximidamide

  • MDL number MFCD00460145
  • Cas Number : 65695-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104263

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104263
Availability details for
Quantity
  • Availability
  • Add to Cart