ENA370563527 Enamine

3-(2-chloro-4-fluorophenyl)-1,2,4-oxadiazole-5-thiol

  • MDL number MFCD17259867
  • Cas Number : 1343988-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104264
Availability details for
Quantity
  • Availability
  • Add to Cart