ENA370563528 Enamine

1-benzyl-5-methylpyrrolidin-3-amine

  • MDL number MFCD20722120
  • Cas Number : 74880-19-6
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzyl-5-methyl-3-pyrrolidinamine
Inchi Code 1S/C12H18N2/c1-10-7-12(13)9-14(10)8-11-5-3-2-4-6-11/h2-6,10,12H,7-9,13H2,1H3
Inchi Key GEZSNOKHPRRTIC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-106080

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-106080
pictograms GHS05,GHS07
hstatements H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart