ENA370563530 Enamine

7-chloro-2,3-dihydro-1-benzofuran-3-amine hydrochloride

  • MDL number MFCD08544180
  • Cas Number : 1384265-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231...233
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106085

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106085
Availability details for
Quantity
  • Availability
  • Add to Cart