ENA370563531 Enamine

methyl({[1-methyl-3-(propan-2-yl)-1H-pyrazol-4-yl]methyl})amine

  • MDL number MFCD12048894
  • Cas Number : 1158052-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106086

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106086
Availability details for
Quantity
  • Availability
  • Add to Cart