ENA370563534 Enamine

1-(4-chlorophenyl)-2,2-dimethylpropan-1-one

  • MDL number MFCD03841177
  • Cas Number : 30314-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106089
Availability details for
Quantity
  • Availability
  • Add to Cart