ENA370563538 Enamine

1-oxaspiro[2.3]hexane

  • MDL number MFCD11226850
  • Cas Number : 157-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106093
Availability details for
Quantity
  • Availability
  • Add to Cart