ENA370563546 Enamine

methyl[2-(4H-1,2,4-triazol-4-yl)ethyl]amine

  • MDL number MFCD16837769
  • Cas Number : 1247663-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106147
Availability details for
Quantity
  • Availability
  • Add to Cart