ENA370563549 Enamine

methyl 2-[(4-nitronaphthalen-1-yl)oxy]acetate

  • MDL number MFCD18998448
  • Cas Number : 874133-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-106163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106163
Availability details for
Quantity
  • Availability
  • Add to Cart