ENA370563550 Enamine

1-(1H-1,2,3-benzotriazole-5-carbonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD12570993

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106164
Availability details for
Quantity
  • Availability
  • Add to Cart