ENA370563551 Enamine

4-[(1H-1,2,3-benzotriazol-5-yl)formamido]butanoic acid

  • MDL number MFCD11549773
  • Cas Number : 1098365-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106167
Availability details for
Quantity
  • Availability
  • Add to Cart