ENA370563552 Enamine

N-(2-bromophenyl)pyrrolidine-1-sulfonamide

  • MDL number MFCD11171792
  • Cas Number : 1042644-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106169

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-106169
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart