ENA370563553 Enamine

5-(2-methoxyethoxy)-1H-indole

  • MDL number MFCD20662719
  • Cas Number : 947380-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106170
Availability details for
Quantity
  • Availability
  • Add to Cart