ENA370563558 Enamine

2-chloro-6-methylpyrimidin-4-amine

  • MDL number MFCD05863311
  • Cas Number : 14394-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220...222
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106183

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106183
Availability details for
Quantity
  • Availability
  • Add to Cart