ENA370563563 Enamine

2-amino-1-(1-benzothiophen-2-yl)ethan-1-ol

  • MDL number MFCD04038403
  • Cas Number : 688763-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106272

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106272
Availability details for
Quantity
  • Availability
  • Add to Cart