ENA370563564 Enamine

4-(1-benzothiophen-3-yl)-4-oxobutanoic acid

  • MDL number MFCD01971126
  • Cas Number : 63471-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101433

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101433
Availability details for
Quantity
  • Availability
  • Add to Cart