ENA370563569 Enamine

2-(2-methylpropyl)aniline

  • MDL number MFCD00130034
  • Cas Number : 71182-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106284
Availability details for
Quantity
  • Availability
  • Add to Cart