ENA370563573 Enamine

4-methoxycyclohexan-1-ol

  • MDL number MFCD00019333
  • Cas Number : 18068-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-106288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106288
Availability details for
Quantity
  • Availability
  • Add to Cart