ENA370563576 Enamine

N-(2-aminophenyl)-N-methylmethanesulfonamide

  • MDL number MFCD16158909
  • Cas Number : 90140-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106291

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106291
Availability details for
Quantity
  • Availability
  • Add to Cart