ENA370563577 Enamine

3-chloro-6-(2-methyl-1H-imidazol-1-yl)pyridazine

  • MDL number MFCD09879090
  • Cas Number : 75792-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104274
Availability details for
Quantity
  • Availability
  • Add to Cart