ENA370563578 Enamine

2,3-dichloropyrido[2,3-b]pyrazine

  • MDL number MFCD09031107
  • Cas Number : 25710-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106294

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-106294
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart