ENA370563579 Enamine

7-chloro-5-phenyl-1,3-benzoxazol-2-amine

  • MDL number MFCD17335538
  • Cas Number : 1274551-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237...239
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104275

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104275
Availability details for
Quantity
  • Availability
  • Add to Cart