ENA370563582 Enamine

[2-(cyclopropylmethoxy)ethyl](methyl)amine

  • MDL number MFCD06246780
  • Cas Number : 883526-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104276

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104276
Availability details for
Quantity
  • Availability
  • Add to Cart