ENA370563584 Enamine

1-cyano-3-methylcyclobutane-1-carboxylic acid

  • MDL number MFCD19683133
  • Cas Number : 1343155-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106305
Availability details for
Quantity
  • Availability
  • Add to Cart