ENA370563585 Enamine

1-(1-benzylcyclobutyl)methanamine hydrochloride

  • MDL number MFCD22369985
  • Cas Number : 1384264-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106312
Availability details for
Quantity
  • Availability
  • Add to Cart